siRNA Details
Gene Symbol: Ift27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025931.2 | NP_080207.1 | 5 | 632 | ||
GenBank mRNA | AK011196.1 | - | 5 | 630 | ||
AK044278.1 | - | 2 | 1070 | |||
BC017514.1 | - | 5 | 632 | 4502942 |
