siRNA Details
Gene Symbol: Syap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025932.2 | NP_080208.2 | 8 | 1078 | ||
GenBank mRNA | AK008390.1 | - | 8 | 1047 | ||
AK014893.1 | - | 8 | 1064 | |||
AK051251.1 | - | 8 | 1035 | |||
AK147055.1 | - | 8 | 1014 | |||
AK154843.1 | - | 8 | 1035 | |||
BC021373.1 | - | 8 | 1008 | 5039534 |
