siRNA Details
Gene Symbol: Tbc1d7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252639.1 | NP_001239568.1 | 7 | 913 | ||
NM_025935.3 | NP_080211.1 | 7 | 893 | |||
GenBank mRNA | AK003271.1 | - | 3 | 292 | ||
AK011356.1 | - | 7 | 752 | |||
AK156543.1 | - | 7 | 821 | |||
BC125307.1 | - | 7 | 740 | 40129822 | ||
BC125309.1 | - | 7 | 740 | 40129824 | ||
BC145157.1 | - | 7 | 760 | 9053708 | ||
DN174647.1 | - | 4 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
