siRNA Details
Gene Symbol: Yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024189.6 | NP_077151.3 | 4 | 522 | ||
GenBank mRNA | AB100452.1 | - | 4 | 512 | ||
AB100453.1 | - | 6 | 581 | |||
AK005008.1 | - | 4 | 506 | |||
AK011702.1 | - | 5 | 593 | |||
AK012787.1 | - | 4 | 428 | |||
AK148634.1 | - | 4 | 466 | |||
AK179353.1 | - | 2 | 219 | |||
AK184096.1 | - | 4 | 307 | |||
BC002192.1 | - | 4 | 395 | 3488691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
