siRNA Details
Gene Symbol: Tmem246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025944.3 | NP_080220.2 | 2 | 982 | ||
GenBank mRNA | AK013226.1 | - | 2 | 911 | ||
AK038514.1 | - | 2 | 921 | |||
AK039788.1 | - | 2 | 916 | |||
AK042679.1 | - | 2 | 989 | |||
AK045523.1 | - | 2 | 836 | |||
AK075994.1 | - | 2 | 866 | |||
AK165526.1 | - | 2 | 921 | |||
BC013800.1 | - | 2 | 827 | 2650271 |
