siRNA Details
Gene Symbol: Romo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164216.1 | NP_001157688.1 | 3 | 419 | ||
NM_001164217.2 | NP_001157689.1 | 3 | 433 | |||
NM_025946.6 | NP_080222.1 | 2 | 517 | |||
GenBank mRNA | AA058111.1 | - | 3 | 419 | ||
AK003488.1 | - | 3 | 328 | |||
AK008300.1 | - | 3 | 354 | |||
AY028425.1 | - | 2 | 412 | |||
BC028749.1 | - | 3 | 314 | 1196408 | ||
CJ221444.1 | - | 3 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
