siRNA Details
Gene Symbol: 1700011H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025956.4 | NP_080232.3 | 4 | 447 | ||
GenBank mRNA | AK005866.1 | - | 4 | 414 | ||
AK007176.1 | - | 4 | 448 | |||
AK165498.1 | - | 3 | 205 | |||
BC026534.1 | - | 4 | 420 | 5042540 |
