siRNA Details
Gene Symbol: Ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141930.1 | NP_001135402.1 | 2 | 184 | ||
NM_023465.4 | NP_075954.1 | 3 | 290 | |||
GenBank mRNA | AB021261.1 | - | 3 | 207 | ||
AK003595.1 | - | 2 | 186 | |||
AK009066.1 | - | 2 | 111 | |||
AK036181.1 | - | 3 | 291 | |||
AK048258.1 | - | 3 | 288 | |||
BC018436.1 | - | 3 | 252 | |||
BC038253.1 | - | 3 | 266 | 4974951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
