siRNA Details
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175114.3 | NP_780323.2 | 12 | 1509 | ||
GenBank mRNA | AK029123.1 | - | 13 | 1907 | ||
AK041436.1 | - | 13 | 1877 | |||
AK122426.1 | - | 13 | 1599 | |||
BC058971.1 | - | 12 | 1451 | 6832507 | ||
BC116262.1 | - | 12 | 1476 | |||
BC116263.1 | - | 12 | 1476 | |||
BC129961.1 | - | 12 | 1476 | |||
BC129962.1 | - | 12 | 1476 |
