siRNA Details
Gene Symbol: D16Ertd472e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252438.1 | NP_001239367.1 | 2 | 561 | ||
NM_001252439.1 | NP_001239368.1 | 2 | 328 | |||
NM_001252440.1 | NP_001239369.1 | 2 | 293 | |||
NM_025967.4 | NP_080243.3 | 2 | 361 | |||
GenBank mRNA | AK005829.1 | - | 2 | 272 | ||
AK009258.1 | - | 2 | 532 | |||
AK044670.1 | - | 2 | 362 | |||
BC019957.1 | - | 2 | 252 | 3966397 | ||
BG078224.2 | - | 2 | 253 | |||
CN662712.1 | - | 2 | 320 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108312, XM_008768532, XM_008768546, XM_008768531, FQ227934.
Gene Ontology Categories:
