siRNA Details
Gene Symbol: Zbtb8os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025970.3 | NP_080246.1 | 7 | 471 | ||
GenBank mRNA | AK007377.1 | - | 7 | 461 | ||
AK008007.1 | - | 7 | 472 | |||
AK132279.1 | - | 482 | ||||
AY071852.1 | - | 7 | 460 | |||
BC026442.1 | - | 466 | ||||
BC037657.1 | - | 441 | ||||
BC046825.1 | - | 478 | ||||
BC094629.1 | - | 7 | 502 | 3376726 | ||
BC125606.1 | - | 7 | 472 | 40130121 | ||
BC125608.1 | - | 7 | 472 | 40130123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
