siRNA Details
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290502.1 | NP_001277431.1 | 6 | 1046 | ||
NM_025978.4 | NP_080254.2 | 6 | 808 | |||
NM_027619.4 | NP_081895.2 | 6 | 808 | |||
GenBank mRNA | AK012246.1 | - | 6 | 770 | ||
AK015308.1 | - | 6 | 768 | |||
AK017763.1 | - | 4 | 406 | |||
AK031605.1 | - | 6 | 1039 | |||
AK035570.1 | - | 6 | 778 | |||
AK041095.1 | - | 6 | 782 | |||
AK044266.1 | - | 6 | 783 | |||
BC021448.1 | - | 6 | 816 | |||
BC024847.1 | - | 6 | 773 | 5355806 | ||
BC138212.1 | - | 6 | 770 | 8861231 | ||
BC145944.1 | - | 6 | 770 | 40131247 |

More Information
Other Species Targeted

Gene Ontology Categories:
