siRNA Details
Gene Symbol: Ndufa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025987.3 | NP_080263.1 | 2 | 267 | ||
GenBank mRNA | AK002749.1 | - | 2 | 214 | ||
AK010988.1 | - | 2 | 216 | |||
AK012328.1 | - | 2 | 216 | |||
BC048451.1 | - | 2 | 267 | 6773780 |

More Information
Other Species Targeted

Gene Ontology Categories:
