siRNA Details
Gene Symbol: Herc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025992.2 | NP_080268.1 | 16 | 2017 | ||
GenBank mRNA | AK147960.1 | - | 16 | 1940 | ||
AK149648.1 | - | 16 | 2009 | |||
AK161293.1 | - | 4 | 623 |
