siRNA Details
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291155.1 | NP_001278084.1 | 5 | 748 | ||
NM_025996.5 | NP_080272.1 | 5 | 1127 | |||
GenBank mRNA | AK011786.1 | - | 5 | 1127 | ||
AK017699.1 | - | 5 | 693 | |||
AK075873.1 | - | 5 | 745 | |||
AK168329.1 | - | 5 | 682 | |||
BC018278.1 | - | 5 | 678 | 4187356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
