siRNA Details
Gene Symbol: Rnf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025999.3 | NP_080275.2 | 2, 3 | 229 | ||
GenBank mRNA | AF353167.1 | - | 2, 3 | 259 | ||
AK006791.1 | - | 2, 3 | 269 | |||
AK011839.1 | - | 2, 3 | 218 | |||
AK015505.1 | - | 2, 3 | 230 | |||
AK154417.1 | - | 2, 3 | 253 | |||
BC018553.1 | - | 2, 3 | 223 | 3582546 | ||
BY265291.1 | - | 2, 3 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
