siRNA Details
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026001.2 | NP_080277.1 | 10 | 796 | ||
GenBank mRNA | AK011914.1 | - | 10 | 835 | ||
AK082752.1 | - | 10 | 797 | |||
BC047997.1 | - | 10 | 797 | 6510494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
