siRNA Details
Gene Symbol: 2610301B20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026005.3 | NP_080281.3 | 6 | 631 | ||
GenBank mRNA | AK003887.1 | - | 1 | 46 | ||
AK011950.1 | - | 6 | 560 | |||
AK146358.1 | - | 6 | 552 | |||
BC058777.1 | - | 6 | 521 | 5397173 | ||
BM948390.1 | - | 6 | 503 | |||
CX220600.1 | - | 6 | 631 |
