siRNA Details
Gene Symbol: Zmat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026015.2 | NP_080291.1 | 6 | 642 | ||
GenBank mRNA | AK003361.1 | - | 6 | 635 | ||
AK007565.1 | - | 6 | 629 | |||
AK012098.1 | - | 6 | 643 | |||
AK013415.1 | - | 5 | 615 | |||
BC031796.1 | - | 6 | 621 | 4205598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
