siRNA Details
Gene Symbol: Yipf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023311.3 | NP_075800.1 | 2 | 240 | ||
GenBank mRNA | AF140226.1 | - | 1 | 58 | ||
AK007671.1 | - | 2 | 200 | |||
AK010089.1 | - | 2 | 200 | |||
AK151425.1 | - | 2 | 160 | |||
AK151461.1 | - | 2 | 160 | |||
AK152440.1 | - | 2 | 202 | |||
AK153123.1 | - | 2 | 199 | |||
AK160004.1 | - | 2 | 178 | |||
AK167526.1 | - | 2 | 200 | |||
AK167953.1 | - | 2 | 171 | |||
AK172133.1 | - | 2 | 178 | |||
AK181274.1 | - | 1 | 33 | |||
AK183845.1 | - | 1 | 32 | |||
AK205561.1 | - | 1 | 13 | |||
BC003317.1 | - | 2 | 169 | 3155690 | ||
BC055301.1 | - | 1 | 151 | 6466806 | ||
BY286385.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
