siRNA Details
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026017.2 | NP_080293.1 | 3, 4 | 611 | ||
GenBank mRNA | AK076150.1 | - | 3, 4 | 625 | ||
AK164602.1 | - | 3, 4 | 553 | |||
BC018265.1 | - | 3, 4 | 611 | 4486464 |

More Information
Other Species Targeted

Gene Ontology Categories:
