siRNA Details
Gene Symbol: Zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026021.3 | NP_080297.1 | 5 | 721 | ||
GenBank mRNA | AJ409150.1 | - | 5 | 682 | ||
AJ409151.1 | - | 5 | 567 | |||
AK005908.1 | - | 5 | 709 | |||
AK012484.1 | - | 5 | 722 | |||
AK088063.1 | - | 5 | 742 | |||
AK184476.1 | - | 1 | 85 | |||
BC027321.1 | - | 5 | 687 | 3990966 |

More Information
Other Species Targeted

Gene Ontology Categories:
