siRNA Details
Gene Symbol: Ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278115.1 | NP_001265044.1 | 6 | 520 | ||
NM_026024.3 | NP_080300.1 | 6 | 697 | |||
GenBank mRNA | AK012565.1 | - | 6 | 656 | ||
AK021213.1 | - | 6 | 686 | |||
AK166098.1 | - | 6 | 668 | |||
BC029213.1 | - | 6 | 646 | 3591557 | ||
CK334733.1 | - | 6 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
