siRNA Details
Gene Symbol: Lsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026032.1 | NP_080308.1 | 3 | 254 | ||
GenBank mRNA | AK041290.1 | - | 3 | 254 | ||
AK048497.1 | - | 3 | 287 | |||
BC021460.1 | - | 3 | 537 | 5054997 |

More Information
Other Species Targeted

Gene Ontology Categories:
