siRNA Details
Gene Symbol: MAGOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002370.3 | NP_002361.1 | 3 | 365 | ||
GenBank mRNA | AF035940.1 | - | 3 | 268 | ||
AF067173.1 | - | 3 | 291 | |||
AK312113.1 | - | 3 | 301 | |||
BC018211.1 | - | 3 | 277 | 3861094 | ||
BX283982.1 | - | 3 | 365 |
