siRNA Details
Gene Symbol: L3hypdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026038.2 | NP_080314.1 | 2 | 743 | ||
GenBank mRNA | AK018449.1 | - | 2 | 744 | ||
AK137154.1 | - | 2 | 831 | |||
BC004753.1 | - | 2 | 803 | 3584353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
