siRNA Details
Gene Symbol: Med29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026042.3 | NP_080318.2 | 2 | 284 | ||
GenBank mRNA | AK005112.1 | - | 2 | 286 | ||
AK013006.1 | - | 2 | 284 | |||
BC023192.1 | - | 2 | 267 | 4947722 | ||
BC083314.1 | - | 2 | 266 | 6823222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
