siRNA Details
Gene Symbol: Zkscan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023322.2 | NP_075811.2 | 3 | 831 | ||
GenBank mRNA | AK012630.1 | - | 2 | 195 | ||
AK013258.1 | - | 3 | 819 | |||
AK153972.1 | - | 3 | 814 | |||
BC141244.1 | - | 3 | 757 | 9055856 | ||
U62906.1 | - | 3 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
