siRNA Details
Gene Symbol: Cinp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026048.4 | NP_080324.1 | 3, 4 | 382 | ||
GenBank mRNA | AK004355.1 | - | 3, 4 | 337 | ||
AK076126.1 | - | 3, 4 | 352 | |||
AK160662.1 | - | 3, 4 | 352 | |||
BC011066.1 | - | 3, 4 | 352 | 3985202 |

More Information
Other Species Targeted

Gene Ontology Categories:
