siRNA Details
Gene Symbol: Cinp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026048.4 | NP_080324.1 | 4 | 429 | ||
NM_027223.1 | NP_081499.1 | 5 | 579 | |||
GenBank mRNA | AK004355.1 | - | 4 | 384 | ||
AK076126.1 | - | 4 | 399 | |||
AK160662.1 | - | 4 | 399 | |||
BC011066.1 | - | 4 | 399 | 3985202 | ||
BC021867.1 | - | 5 | 531 | 3710025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
