siRNA Details
Gene Symbol: 2810474O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 2777 | ||
NM_001289662.1 | NP_001276591.1 | 3 | 2690 | |||
NM_026054.3 | NP_080330.1 | 3 | 2708 | |||
GenBank mRNA | AK031526.1 | - | 3 | 2571 | ||
AK031989.1 | - | 4 | 2740 | |||
AK079405.1 | - | 4 | 2608 | |||
AK220403.1 | - | 3 | 2691 | |||
BC023168.1 | - | 1 | 684 | |||
BC086679.1 | - | 1 | 2128 | |||
BC139435.1 | - | 3 | 2630 | 8862457 | ||
BC139436.1 | - | 4 | 2717 | 8862458 |
