siRNA Details
Gene Symbol: Rpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026055.1 | NP_080331.1 | 2, 3 | 170 | ||
GenBank mRNA | AK002591.1 | - | 2, 3 | 168 | ||
AK005511.1 | - | 2, 3 | 170 | |||
AK007821.1 | - | 2, 3 | 172 | |||
AK008853.1 | - | 2, 3 | 168 | |||
AK013381.1 | - | 2, 3 | 174 | |||
AK028139.1 | - | 2, 3 | 170 | |||
AK088761.1 | - | 2, 3 | 175 | |||
AK146772.1 | - | 2, 3 | 172 | |||
AK150955.1 | - | 2, 3 | 141 | |||
AK168773.1 | - | 2, 3 | 168 | |||
BC039092.1 | - | 2, 3 | 145 | 5374136 | ||
BC094645.1 | - | 2, 3 | 151 | 1532664 | ||
BC139247.1 | - | 2, 3 | 145 | 8862269 | ||
BC139249.1 | - | 2, 3 | 146 | 8862271 |

More Information
Other Species Targeted

Gene Ontology Categories:
