siRNA Details
Gene Symbol: 2900011O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144518.3 | NP_653101.1 | 5 | 617 | ||
GenBank mRNA | AK035868.1 | - | 5 | 594 | ||
AK049997.1 | - | 5 | 649 | |||
AK076136.1 | - | 5 | 636 | |||
AK136859.1 | - | 5 | 617 | |||
AK139470.1 | - | 5 | 629 | |||
BC022741.1 | - | 5 | 596 | 4511454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
