siRNA Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 10, 11 | 1188 | ||
GenBank mRNA | AK013554.1 | - | 10, 11 | 1140 | ||
AK021316.1 | - | 3, 4 | 295 | |||
AK028937.1 | - | 10, 11 | 1114 | |||
AK028945.1 | - | 10, 11 | 1091 | |||
AK030280.1 | - | 10, 11 | 1128 | |||
AK076409.1 | - | 1, 2 | 122 | |||
AK134887.1 | - | 9, 10 | 1847 | |||
AK210387.1 | - | 1, 2 | 142 | |||
AY029531.1 | - | 10, 11 | 1089 | |||
BC003946.1 | - | 10, 11 | 1075 | 3487343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
