siRNA Details
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024197.1 | NP_077159.1 | 8, 9 | 917 | ||
GenBank mRNA | AK005339.1 | - | 8, 9 | 953 | ||
AK013675.1 | - | 8, 9 | 960 | |||
AK046292.1 | - | 821 | ||||
AK145979.1 | - | 8, 9 | 973 | |||
BC003439.1 | - | 8, 9 | 917 | 2647815 |
