siRNA Details
Gene Symbol: Med31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026068.2 | NP_080344.2 | 4 | 820 | ||
GenBank mRNA | AK005655.1 | - | 4 | 229 | ||
AK013990.1 | - | 4 | 261 | |||
BC019685.1 | - | 4 | 290 | 3983617 | ||
BC051929.1 | - | 4 | 820 | 5709831 |
