siRNA Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122960.1 | NP_001116432.1 | 4 | 409 | ||
NM_026070.3 | NP_080346.1 | 4 | 409 | |||
NM_027487.1 | NP_081763.1 | 4 | 409 | |||
GenBank mRNA | AK004206.1 | - | 4 | 324 | ||
AK009358.1 | - | 4 | 305 | |||
AK013835.1 | - | 4 | 414 | |||
AK017732.1 | - | 4 | 337 | |||
AK158822.1 | - | 4 | 410 | |||
BC029256.1 | - | 4 | 306 | 3661472 | ||
CK795584.1 | - | 4 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
