siRNA Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026070.3 | NP_080346.1 | 4, 5 | 474 | ||
NM_027487.1 | NP_081763.1 | 4, 5 | 474 | |||
GenBank mRNA | AK004206.1 | - | 4, 5 | 389 | ||
AK009358.1 | - | 4, 5 | 370 | |||
AK013835.1 | - | 4, 5 | 479 | |||
AK017732.1 | - | 4, 5 | 402 | |||
AK158822.1 | - | 4, 5 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
