siRNA Details
Gene Symbol: Cltc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003908.1 | NP_001003908.1 | 3 | 564 | ||
GenBank mRNA | AK122193.1 | - | 3 | 568 | ||
AK156869.1 | - | 3 | 583 | |||
AK158438.1 | - | 3 | 550 | |||
AK171732.1 | - | 3 | 566 | |||
BC079897.1 | - | 3 | 564 | 30546407 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019299, XM_006247107, XM_008768108, XM_006247109, XM_006247108, and others.
Gene Ontology Categories:
