siRNA Details
Gene Symbol: Gpx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024198.3 | NP_077160.1 | 2 | 325 | ||
GenBank mRNA | AK004091.1 | - | 2 | 318 | ||
AK014199.1 | - | 2 | 323 | |||
AK084382.1 | - | 2 | 328 | |||
AK165221.1 | - | 2 | 326 | |||
BC003228.1 | - | 2 | 326 | 3483448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
