siRNA Details
Gene Symbol: Prl8a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023332.3 | NP_075821.1 | 6 | 711 | ||
GenBank mRNA | AF158744.1 | - | 6 | 696 | ||
AK005409.1 | - | 6 | 712 | |||
AK005494.1 | - | 6 | 712 | |||
BC064762.1 | - | 6 | 679 | 30013931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
