siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026101.4 | NP_080377.2 | 12, 13 | 1816 | ||
NM_030114.2 | NP_084390.1 | 12, 13 | 1816 | |||
GenBank mRNA | AK019546.1 | - | 12, 13 | 1812 | ||
AK143111.1 | - | 12, 13 | 1690 | |||
AK145735.1 | - | 7, 8 | 764 | |||
AK147723.1 | - | 12, 13 | 1703 | |||
BC043082.1 | - | 12, 13 | 1628 | 6417111 | ||
BC060033.1 | - | 10, 11 | 1401 | 6478123 |
