siRNA Details
Gene Symbol: 1700093K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110133.1 | NP_001103603.1 | 2 | 565 | ||
NM_026105.3 | NP_080381.2 | 2 | 247 | |||
GenBank mRNA | AK007052.1 | - | 2 | 153 | ||
BC049567.1 | - | 2 | 247 | 6773879 | ||
CF578320.1 | - | 2 | 593 |
