siRNA Details
Gene Symbol: Gtf3c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026113.4 | NP_080389.2 | 6 | 412 | ||
GenBank mRNA | AK005261.1 | - | 6 | 411 | ||
AK007816.1 | - | 6 | 413 | |||
AK010514.1 | - | 6 | 412 | |||
AK162053.1 | - | 6 | 412 | |||
AK168070.1 | - | 6 | 411 | |||
AK185766.1 | - | 2 | 121 | |||
BC027260.1 | - | 5 | 328 | |||
BC048446.1 | - | 6 | 418 | 6590885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
