siRNA Details
Gene Symbol: Qprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133686.1 | NP_598447.1 | 4 | 1151 | ||
GenBank mRNA | AK010602.1 | - | 4 | 1165 | ||
AK019119.1 | - | 4 | 1166 | |||
BC011191.1 | - | 4 | 1151 | 4196244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
