siRNA Details
Gene Symbol: Bbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026116.2 | NP_080392.1 | 14, 15 | 2264 | ||
GenBank mRNA | AF342737.1 | - | 14, 15 | 2264 | ||
AK010779.1 | - | 14, 15 | 1877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
