siRNA Details
Gene Symbol: Med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026119.3 | NP_080395.1 | 2 | 160 | ||
GenBank mRNA | AK010270.1 | - | 2 | 162 | ||
AK010686.1 | - | 2 | 162 | |||
AK078288.1 | - | 2 | 172 | |||
AK145636.1 | - | 2 | 185 | |||
BC024950.1 | - | 2 | 160 | 3992287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
