siRNA Details
Gene Symbol: 1110008F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026124.3 | NP_080400.1 | 3 | 442 | ||
GenBank mRNA | AK002452.1 | - | 3 | 400 | ||
AK003568.1 | - | 3 | 437 | |||
AK179243.1 | - | 3 | 163 | |||
AK186989.1 | - | 3 | 163 | |||
BC019445.1 | - | 3 | 380 | 5125733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
