siRNA Details
Gene Symbol: 1110008F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026124.3 | NP_080400.1 | 3 | 450 | ||
GenBank mRNA | AK002452.1 | - | 3 | 408 | ||
AK003568.1 | - | 3 | 445 | |||
AK179243.1 | - | 3 | 171 | |||
BC019445.1 | - | 3 | 388 | 5125733 |
