siRNA Details
Gene Symbol: Fam132a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026125.3 | NP_080401.1 | 3 | 647 | ||
GenBank mRNA | AK002876.1 | - | 3 | 629 | ||
AK003510.1 | - | 3 | 407 | |||
AK004116.1 | - | 3 | 642 | |||
AK007335.1 | - | 3 | 632 | |||
BC026939.1 | - | 3 | 619 | 3966551 | ||
DQ002404.1 | - | 3 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
